Fast Plus EvaGreen®, qPCR Master Mixes, Biotium
Supplier: Biotium
31014 31014-1 31014-2 31014-T 31015 31015-1 31015-2 31015-T 31020-1 31020-2 31020-T 31020
89139-026EA 215 USD
89139-026 89139-028 89139-030 89139-032 89139-034 89139-036 89139-038 89139-040 89260-212 89260-214 89260-216 89410-662
Fast Plus EvaGreen®, qPCR Master Mixes, Biotium
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
A qPCR master mix containing Evagreen® qPCR dye and Cheetah™ Taq hotstart DNA polymerase, suitable for qPCR using a fast cycling protocol.
  • 2X qPCR master mix for fast cycling protocols
  • EvaGreen® dye-based
  • Contains Cheetah™ Taq hotstart DNA polymerase
Fast Plus EvaGreen® qPCR Master Mix contains Evagreen® qPCR dye and Cheetah™ Taq hotstart DNA polymerase. It is suitable for qPCR using a fast cycling protocol. However for most PCR applications we recommend the Forget-Me-Not™ series of master mixes, for superior activity.
Fast Plus Evagreen® qPCR Master Mix is a 2X master mix, and is supplied with a separate vial of ROX reference dye, for use with high ROX, low ROX, or no ROX instruments.