Anti-FMC63 Rabbit Polyclonal Antibody

Supplier: ACRO Biosystems
FM3S931MG FM3S93100UG
103889-738EA 3258 USD
103889-738 103889-740
Anti-FMC63 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FMC63
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Polyclonal
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR