Anti-Spike RBD HEK293 Monoclonal Antibody (Precision Avi)

Supplier: ACRO Biosystems
S1NVM226200UG S1NVM22625UG
103887-952EA 2160 USD
103887-952 103887-954
Anti-Spike RBD HEK293 Monoclonal Antibody (Precision Avi)
Antibodies
Type: Primary
Antigen: Spike RBD
Clonality: Monoclonal
Clone:
Conjugation: Precision Avi
Epitope:
Host: HEK293
Isotype: Human IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR