2-Deoxy-D(-)-ribose 98%

Supplier: Ambeed
Warning

A358856-100G A358856-500G A358856-25G A358856-5G
103837-220EA 9 USD
103837-220 77278-256 77278-258 77306-696 77306-698
2-Deoxy-D(-)-ribose 98%
2-Deoxy-D(-)-ribose

Caution: for Research Use Only

Formula: C₅H₁₀O₄
Boiling Pt: 379.7 °C (760 mm Hg)
Melting Pt: 89…90 °C
Flash Pt: 154.6 °C
Storage Temperature: Freezer
MDL Number: MFCD00135904
CAS Number: 533-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR