3-Amino-1-Boc-piperidine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-BOC-3-Aminopiperidine, (±)-3-Amino-1-boc-piperidine, tert-Butyl 3-aminopiperidine-1-carboxylate
A145731-100G
A145731-10G
A145731-25G
A145731-1G
A145731-5G
103803-574EA
9
USD
103803-574
103841-656
76767-976
76767-978
76767-980
77316-960
77316-962
3-Amino-1-Boc-piperidine 97%
3-Amino-1-Boc-piperidine
Caution: for Research Use Only
Formula:
C₁₀H₂₀N₂O₂ MW: 200.28 g/mol Boiling Pt: 277.3 °C (760 mm Hg) Melting Pt: 181…182 °C Flash Pt: 121.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01861219 CAS Number: 184637-48-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...