Boc-Aib-ol 96%

Supplier: Ambeed
Warning

Synonyms: N-Boc-2-amino-2-methyl-1-propanol, N-tert-Butoxycarbonyl-2-amino-2-methyl-1-propanol, tert-Butyl (1-Hydroxy-2-methylpropan-2-yl)carbamate

A263074-100G A263074-10G A263074-25G A263074-5G A263074-1G
103812-604EA 7 USD
103812-604 103852-500 76977-018 76977-020 77092-130 77092-132 77304-866
Boc-Aib-ol 96%
Boc-Aib-ol

Caution: for Research Use Only

Formula: C₉H₁₉NO₃
MW: 189.3 g/mol
Melting Pt: 57…62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03788641
CAS Number: 102520-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR