1,9-Pyrazoloanthrone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
JNK Inhibitor II, Anthra[1,9-cd]pyrazol-6(2H)-one, Dibenzo[cd,g]indazol-6(2H)-one, SP600125, SP 600125
A155219-100MG
A155219-10MG
A155219-50MG
A155219-1MG
103804-734EA
94
USD
103804-734
103842-928
76783-542
76783-544
77549-970
77600-214
1,9-Pyrazoloanthrone 98%
1,9-Pyrazoloanthrone
Caution: for Research Use Only
Formula:
C₁₄H₈N₂O MW: 220.231 g/mol Boiling Pt: 489.3 °C (760 mm Hg) Melting Pt: 281…282 °C Flash Pt: 246.8 °C Storage Temperature: Freezer |
MDL Number:
MFCD00022289 CAS Number: 129-56-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...