1-Octanesulfonic acid sodium salt 98%

Supplier: Ambeed
Warning

Synonyms: Sodium 1-octansulfonate, Sodium octane-1-sulphonate

A338599-5G A338599-100G A338599-10G A338599-25G A338599-500G
103836-742EA 9 USD
103836-742 76839-766 77239-840 77239-842 77306-512 77593-976
1-Octanesulfonic acid sodium salt 98%
1-Octanesulphonic acid sodium salt

Caution: for Research Use Only

Formula: C₈H₁₇NaO₃S
MW: 216.28 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007544
CAS Number: 5324-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR