HEPES sodium salt 98%

Supplier: Ambeed
Warning

Synonyms: Sodium 4-(2-hydroxyethyl)piperazin-1-ylethanesulphonate, 4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, Sodium 2-(4-(2-hydroxyethyl)piperazin-1-yl)ethanesulphonate, Sodium 2-(4-(2-hydroxyethyl)piperazin-1-yl)ethanesulfonate, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid) sodium salt, 4-(2-Hydroxyethyl)piperazine-1-ethanesulphonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid) sodium salt, HEPES sodium, Sodium 4-(2-hydroxyethyl)piperazin-1-ylethanesulfonate, HEPES Na

A260290-100G A260290-1000G A260290-25G A260290-500G
103809-850EA 144 USD
103809-850 77031-744 77304-784 77304-786 77672-322
HEPES sodium salt 98%
HEPES sodium salt

Caution: for Research Use Only

Formula: C₈H₁₇N₂NaO₄S
MW: 260.29 g/mol
Melting Pt: 234 °C
Storage Temperature: Ambient
MDL Number: MFCD00036463
CAS Number: 75277-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR