Silver carbonate 98%

Supplier: Ambeed
Danger

A471802-10G A471802-1G A471802-25G A471802-5G A471802-100G A471802-500G
103828-032EA 6 USD
103828-032 103839-106 76725-798 77307-484 77307-486 77307-488 77572-484 77872-785
Silver carbonate 98%
Silver carbonate

Caution: for Research Use Only

Formula: CAg₂O₃
Boiling Pt: 333.6 °C (760 mm Hg)
Melting Pt: 210 °C
Storage Temperature: Ambient
MDL Number: MFCD00003403
CAS Number: 534-16-7
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR