Bisindolylmaleimide IX·methanesulfonate 98%

Supplier: Ambeed
Warning

Synonyms: Ro 31-8220 mesylate

A350912-10MG A350912-1MG A350912-50MG A350912-5MG
103791-648EA 31 USD
103791-648 103795-140 103799-618 76724-464 76840-144 76840-146 76840-148
Bisindolylmaleimide IX·methanesulfonate 98%
Bisindolylmaleimide IX·methanesulfonate

Caution: for Research Use Only

Formula: C₂₅H₂₃N₅O₂S·CH₄O₃S
Storage Temperature: Freezer
MDL Number: MFCD03788207
CAS Number: 138489-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR