Resiquimod 98%

Supplier: Ambeed
Warning

Synonyms: 1-[4-Amino-2-(ethoxymethyl)-1H-imidazo[4,5-c]quinolin-1-yl]-2-methylpropan-2-ol, 4-Amino-2-(ethoxymethyl)-a,a-dimethyl-1H-imidazo[4,5-c]quinoline-1-ethanol, 1-(4-Amino-2-(ethoxymethyl)-1H-imidazo[4,5-c]quinolin-1-yl)-2-methylpropan-2-ol, 4-Amino-2-(ethoxymethyl)-alpha,alpha-dimethyl-1H-imidazo[4,5-c]quinoline-1-ethanol, R-848

A119633-100MG A119633-1MG A119633-250MG A119633-5G A119633-1G
103792-066EA 40 USD
103792-066 103792-068 103799-806 103799-306 76810-404 76810-406 76810-408 76810-410 77311-916
Resiquimod 98%
Resiquimod

Caution: for Research Use Only

Formula: C₁₇H₂₂N₄O₂
Boiling Pt: 553.6 °C (760 mm Hg)
Melting Pt: 166 °C
Flash Pt: 288.6 °C
Storage Temperature: Freezer
MDL Number: MFCD00937759
CAS Number: 144875-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR