Nortropinone hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
8-Azabicyclo[3.2.1]octan-3-one hydrochloride
A102881-100G
A102881-10G
A102881-25G
A102881-500G
A102881-5G
103822-138EA
5
USD
103822-138
77093-580
77093-582
77093-584
77093-586
77309-010
Nortropinone hydrochloride 97%
Nortropinone hydrochloride
Caution: for Research Use Only
Formula:
C₇H₁₂ClNO MW: 161.63 g/mol Boiling Pt: 258.8 °C (760 mm Hg) Flash Pt: 110.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD03613582 CAS Number: 25602-68-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...