Nobiletin 98%

Supplier: Ambeed
Warning

Synonyms: 3',4',5,6,7,8-Hexamethoxyflavone, 2-(3,4-Dimethoxyphenyl)-5,6,7,8-tetramethoxy-4H-chromen-4-one

A298598-1MG A298598-250MG
103791-616EA 18 USD
103791-616 103792-574 103795-120 103799-580 77305-744 77748-580
Nobiletin 98%
Nobiletin

Caution: for Research Use Only

Formula: C₂₁H₂₂O₈
MW: 402.40 g/mol
Boiling Pt: 587.9 °C (760 mm Hg)
Flash Pt: 256.5 °C
Storage Temperature: Freezer
MDL Number: MFCD03273560
CAS Number: 478-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR