2,6-Dihydroxynaphthalene 98%

Supplier: Ambeed
Warning

Synonyms: Naphthalene-2,6-diol

A108808-100G A108808-10G A108808-25G A108808-500G A108808-1G A108808-5G
103797-390EA 6 USD
103797-390 103823-180 77107-844 77107-846 77107-848 77107-850 77107-852 77310-006 77310-008
2,6-Dihydroxynaphthalene 98%
2,6-Dihydroxynaphthalene

Caution: for Research Use Only

Formula: C₁₀H₈O₂
MW: 160.17 g/mol
Boiling Pt: 375
Melting Pt: 220
Flash Pt: 193.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00004082
CAS Number: 581-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR