N,N'-Dicyclohexylmorpholine-4-carboximidamide 95%
Supplier: Ambeed
|
Warning
|
A281760-5G
A281760-10G
A281760-25G
103853-520EA
22
USD
103853-520
76837-218
76977-434
77305-414
N,N'-Dicyclohexylmorpholine-4-carboximidamide 95%
N,N'-Dicyclohexylmorpholine-4-carboximidamide
Caution: For research use only
Formula:
C₁₇H₃₁N₃O MW: 293.45 g/mol Boiling Pt: 423.2 °C (760 mm Hg) Melting Pt: 105…107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063252 CAS Number: 4975-73-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...