Minoxidil 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,6-Diamino-4-(piperidin-1-yl)pyrimidine 1-oxide, Rogaine, Loniten
A141385-10G
A141385-1G
A141385-1MG
A141385-25G
A141385-100G
103834-022EA
26
USD
103834-022
76719-378
76719-380
76719-382
77316-144
77667-050
Minoxidil 95%
Minoxidil
Caution: for Research Use Only
Formula:
C₉H₁₅N₅O MW: 209.253 g/mol Boiling Pt: 351.7 °C (760 mm Hg) Melting Pt: 272…274 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00063409 CAS Number: 38304-91-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...