Methyl-3-aminobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-aminobenzoate

A142269-100G A142269-10G A142269-500G A142269-25G A142269-5G
103834-168EA 8 USD
103834-168 76972-662 76972-664 76972-666 77316-352 77316-354
Methyl-3-aminobenzoate 98%
Methyl 3-aminobenzoate

Caution: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 120
Melting Pt: 50…52 °C
Flash Pt: 140.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00017102
CAS Number: 4518-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR