Methyl-2-fluorobenzoate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 2-fluorobenzoate, 2-Fluorobenzoic acid emthyl ester
A101960-1000G
A101960-100G
A101960-10G
A101960-25G
A101960-500G
A101960-5G
103821-924EA
5
USD
103821-924
76717-600
76717-602
76717-604
76717-606
76717-608
77282-878
Methyl-2-fluorobenzoate 98%
Methyl 2-fluorobenzoate
Caution: for Research Use Only
Formula:
C₈H₇FO₂ MW: 154.14 g/mol Boiling Pt: 109…110 °C (35 mm Hg) Flash Pt: 93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017913 CAS Number: 394-35-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...