Methyl-2-fluoro-4-nitrobenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-fluoro-4-nitrobenzoate

A429249-1G A429249-25G A429249-5G A429249-10G A429249-250MG
103827-502EA 6 USD
103827-502 103838-558 76842-238 76842-240 76842-242 77033-914 77033-916
Methyl-2-fluoro-4-nitrobenzoate 97%
Methyl 2-fluoro-4-nitrobenzoate

Caution: for Research Use Only

Formula: C₈H₆FNO₄
Boiling Pt: 314.6 °C (760 mm Hg)
Flash Pt: 144.1 °C
Storage Temperature: Ambient
MDL Number: MFCD08444028
CAS Number: 392-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR