Lamivudine 98%

Supplier: Ambeed
Warning

Synonyms: (-)-1-[(2R,5S)-2-(Hydroxymethyl)-1,3-oxathiolan-5-yl]cytosine, 4-Amino-1-((2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl)pyrimidin-2(1H)-one, 3TC

A107352-100G A107352-25G A107352-25MG A107352-500G
103822-946EA 8 USD
103822-946 76995-006 76995-008 76995-010 77549-066
Lamivudine 98%
Lamivudine

Caution: for Research Use Only

Formula: C₈H₁₁N₃O₃S
MW: 229.2600 g/mol
Boiling Pt: 475.4 °C (760 mm Hg)
Melting Pt: 177 °C
Flash Pt: 241.3 °C
Storage Temperature: Freezer
MDL Number: MFCD00869739
CAS Number: 134678-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR