Isoxazolo[4,5-b]pyridin-3-amine 98%
Supplier: Ambeed
|
Warning
|
A141730-100MG
A141730-250MG
103803-052EA
100
USD
103803-052
103834-088
76782-968
76782-970
Isoxazolo[4,5-b]pyridin-3-amine 98%
Isoxazolo[4,5-b]pyridin-3-amine
Caution: for Research Use Only
Formula:
C₆H₅N₃O MW: 135.1200 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13193314 CAS Number: 114080-93-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...