Isonicotinamide 98%

Supplier: Ambeed
Warning

Synonyms: 4-Pyridinecarboxylic acid amide

A154365-100G A154365-500G
103808-798EA 87 USD
103808-798 103842-816 77567-248 77881-209
Isonicotinamide 98%
Isonicotinamide

Caution: For research use only. Inert atmosphere, room temperature

Formula: C₆H₆N₂O
MW: 122.13 g/mol
Boiling Pt: 334
Melting Pt: 155…157 °C
Flash Pt: 156 °C
Storage Temperature: Ambient
MDL Number: MFCD00006432
CAS Number: 1453-82-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR