Hexadecyl palmitate 97%

Supplier: Ambeed
Warning

Synonyms: Cetyl Palmitate, Palmitic acid palmityl ester, n-Hexadecyl palmitate, Palmitic Acid Hexadecyl Ester, Palmityl palmitate

A140387-100G A140387-10G A140387-500G A140387-25G A140387-5G
103833-884EA 6 USD
103833-884 76688-998 76689-000 76689-002 77299-136 77299-138
Hexadecyl palmitate 97%
Hexadecyl palmitate

Caution: for Research Use Only

Formula: C₃₂H₆₄O₂
MW: 480.86 g/mol
Melting Pt: 55…56 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00053739
CAS Number: 540-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR