H-D-Asp(OBzl)-OH 97%

Supplier: Ambeed
Warning

A151353-10G A151353-1G A151353-25G A151353-5G A151353-100G
103804-250EA 8 USD
103804-250 103842-402 76997-350 77317-952 77317-954 77317-956 77881-172
H-D-Asp(OBzl)-OH 97%
H-D-Asp(OBzl)-OH

Caution: for Research Use Only

Formula: C₁₁H₁₃NO₄
MW: 223.228 g/mol
Boiling Pt: 413.1±45.0 °C (760 mm Hg)
Flash Pt: 203.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077100
CAS Number: 13188-89-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR