D-Gluconic acid 50% in water
Supplier: Ambeed
|
Warning
|
Synonyms:
Gluconic acid, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanoic acid
A278252-25G
A278252-100G
A278252-500G
A278252-1000G
103853-346EA
5
USD
103853-346
77093-260
77153-768
77153-770
77685-783
D-Gluconic acid 50% in water
D-Gluconic acid
Caution: for Research Use Only
Formula:
C₆H₁₂O₇ MW: 196.16 g/mol Boiling Pt: 673.6 °C (760 mm Hg) Flash Pt: 375.1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004240 CAS Number: 526-95-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...