Formylhydrazine 98%

Supplier: Ambeed
Warning

Synonyms: Formic acid hydrazide, Formohydrazide, Formic hydrazide

A171403-100G A171403-10G A171403-25G A171403-5G
103844-904EA 9 USD
103844-904 76799-420 76799-422 77082-056 77082-058
Formylhydrazine 98%
Formylhydrazine

Caution: for Research Use Only

Formula: CH₄N₂O
MW: 60.06
Boiling Pt: 90
Melting Pt: 150
Storage Temperature: Freezer
MDL Number: MFCD00007589
CAS Number: 624-84-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR