N-Fmoc-D-glutamic acid 5-tert-butyl ester 97%

Supplier: Ambeed
Warning

Synonyms: 5-tert-Butyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-glutamate, 5-tert-Butyl N-Fmoc-D-glutamate, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-glutamic Acid 5-tert-Butyl Ester, Fmoc-D-Glu(OtBu)-OH, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(tert-butoxy)-5-oxopentanoic acid

A252129-10G A252129-1G A252129-25G A252129-5G A252129-100G
103812-024EA 8 USD
103812-024 103851-868 77031-544 77091-268 77091-270 77091-272 77550-600
N-Fmoc-D-glutamic acid 5-tert-butyl ester 97%
N-Fmoc-D-glutamic acid 5-tert-butyl ester

Caution: for Research Use Only

Formula: C₂₄H₂₇NO₆
MW: 425.48 g/mol
Boiling Pt: 633.5 °C (760 mm Hg)
Melting Pt: 83…89 °C
Flash Pt: 336.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00077055
CAS Number: 104091-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR