Fenofibrate 97%

Supplier: Ambeed
Warning

A763996-25G A763996-100G A763996-1G A763996-250MG A763996-5G A763996-10G A763996-500G
103840-904EA 8 USD
103840-904 76845-136 76981-448 76981-450 76981-452 77308-282 77674-096 77674-098
Fenofibrate 97%
Fenofibrate

Caution: For research use only

Formula: C₂₀H₂₁ClO₄
Storage Temperature: Ambient
MDL Number: MFCD00133314
CAS Number: 49562-28-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR