Ethyl (ethoxymethylene)acetate 98%

Supplier: Ambeed
Danger

Synonyms: 3-Ethoxyacrylic acid ethyl ester, Ethyl 3-Ethoxyacrylate, Ethyl 3-Ethoxy-2-Propenoate, Ethyl 3-ethoxyacrylate

A159605-100G A159605-10G A159605-1G A159605-25G A159605-5G A159605-500G
103843-490EA 6 USD
103843-490 76798-966 76798-968 76798-970 77319-378 77319-380 77667-246
Ethyl (ethoxymethylene)acetate 98%
Ethyl (ethoxymethylene)acetate

Caution: for Research Use Only

Formula: C₇H₁₂O₃
MW: 144.17 g/mol
Boiling Pt: 195.5 °C (760 mm Hg)
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00009863
CAS Number: 1001-26-9
UN: 3334
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR