Ethyl (R)-(-)-3-hydroxybutyrate 96%

Supplier: Ambeed
Warning

Synonyms: Ethyl (R)-3-hydroxybutyrate, (R)-(-)-3-Hydroxybutyric Acid Ethyl Ester

A110143-100G A110143-10G A110143-25G A110143-5G
103797-624EA 5 USD
103797-624 103823-432 77148-662 77148-664 77148-666 77283-942 77310-234
Ethyl (R)-(-)-3-hydroxybutyrate 96%
Ethyl (R)-(-)-3-hydroxybutyrate

Caution: for Research Use Only

Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 175 °C (760 mm Hg)
Flash Pt: 64.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075386
CAS Number: 24915-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR