(R)-(+)-2-Pyrrolidinone-5-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: D-Pyr-OH, (R)-(+)-2-Pyrrolidone-5-carboxylic acid

A167373-25G A167373-100G A167373-10G A167373-5G
103844-438EA 5 USD
103844-438 77081-450 77273-578 77273-580 77300-892
(R)-(+)-2-Pyrrolidinone-5-carboxylic acid 98%
(R)-(+)-2-Pyrrolidinone-5-carboxylic acid

Caution: for Research Use Only

Formula: C₅H₇NO₃
Boiling Pt: 453.1 °C (760 mm Hg)
Melting Pt: 155…162 °C
Flash Pt: 227.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00066212
CAS Number: 4042-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR