2',3'-Dideoxyinosine 98%

Supplier: Ambeed
Warning

Synonyms: 9-((2R,5S)-5-(Hydroxymethyl)tetrahydrofuran-2-yl)-9H-purin-6-ol

A259943-50MG A259943-10MG
103812-470EA 23 USD
103812-470 103852-352 76692-680 77626-354
2',3'-Dideoxyinosine 98%
2',3'-Dideoxyinosine

Caution: for Research Use Only

Formula: C₁₀H₁₂N₄O₃
MW: 236.23 g/mol
Boiling Pt: 586.5 °C (760 mm Hg)
Melting Pt: 193…195 °C
Storage Temperature: Freezer
MDL Number: MFCD00077728
CAS Number: 69655-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR