Dichloro(p-cymene)ruthenium(II) dimer 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Di(μ-chloro)dichlorobis(p-cymene)diruthenium(II)
A611320-250MG
A611320-1G
A611320-10G
A611320-100G
103840-168EA
69
USD
103840-168
76806-064
77307-942
77551-830
77880-310
Dichloro(p-cymene)ruthenium(II) dimer 98%
Dichloro(p-cymene)ruthenium(II) dimer
Caution: for Research Use Only
Formula:
C₂₀H₂₈Cl₄Ru₂ Melting Pt: 247.0…250.0 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064793 CAS Number: 52462-29-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...