D-(+)-Cellobiose 98%

Supplier: Ambeed
Warning

Synonyms: β-D-Glc-(1→4)-D-Glc, 4-β-D-glucopyranosyl-D-glueopyranose, Cellose, β-Cellobiose, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, 4-O-β-D-Glucopyranosyl-D-glucopyranose

A261924-50MG A261924-25G A261924-5G A261924-100G
103852-436EA 20 USD
103852-436 76692-720 77092-052 77304-836 77601-118
D-(+)-Cellobiose 98%
D(+)-Cellobiose

Caution: for Research Use Only

Formula: C₁₂H₂₂O₁₁
MW: 342.30 g/mol
Boiling Pt: 667.931 °C (760 mm Hg)
Melting Pt: 239 °C
Flash Pt: 357.752 °C
Storage Temperature: Ambient
MDL Number: MFCD00136034
CAS Number: 528-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR