Cytarabine 98%

Supplier: Ambeed
Warning

Synonyms: Cytosine beta-D-Arabinofuranoside, 1-β-D-Arabinofuranosylcytosine, Arabinocytidine, Ara-C, 4-Amino-1-((2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one, Cytosine arabinoside, 4-Amino-1-beta-D-arabinofuranosyl-2(1H)-pyrimidinone

A568740-25G A568740-100G A568740-250MG A568740-1G
103828-796EA 19 USD
103828-796 103839-940 76844-070 77280-088 77280-090 77307-842
Cytarabine 98%
Cytarabine

Caution: for Research Use Only

Formula: C₉H₁₃N₃O₅
MW: 243.22 g/mol
Melting Pt: 220 °C
Flash Pt: 283.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00066487
CAS Number: 147-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR