Cyclopentyl phenyl ketone 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Cyclopentyl(phenyl)methanone, Benzoylcyclopentane
A772160-100G
A772160-1G
A772160-25G
103840-940EA
23
USD
103840-940
76652-902
76845-192
76845-194
Cyclopentyl phenyl ketone 95%
Cyclopentyl phenyl ketone
Caution: for Research Use Only
Formula:
C₁₂H₁₄O MW: 174.24 g/mol Boiling Pt: 273.9 °C (760 mm Hg) Flash Pt: 112.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001378 CAS Number: 5422-88-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...