Boc-Orn(Fmoc)-OH 98%

Supplier: Ambeed
Warning

Synonyms: Nα-Boc-Nδ-Fmoc-L-ornithine

A251922-1G A251922-5G A251922-10G A251922-250MG
103812-014EA 18 USD
103812-014 103851-856 77091-254 77091-256 77098-692 77098-694
Boc-Orn(Fmoc)-OH 98%
Boc-Orn(Fmoc)-OH

Caution: for Research Use Only

Formula: C₂₅H₃₀N₂O₆
MW: 454.523 g/mol
Boiling Pt: 679 °C (760 mm Hg)
Melting Pt: 150…154 °C
Flash Pt: 364.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00080279
CAS Number: 150828-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR