9-Anthrealdehyde 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
9-Anthracenecarbaldehyde, 9-Anthracenecarboxaldehyde, 9-Anthraldehyde
A152071-100G
A152071-10G
A152071-500G
A152071-25G
A152071-5G
103842-502EA
5
USD
103842-502
76812-252
76812-254
76812-256
77318-110
77318-112
9-Anthrealdehyde 98%
9-Anthrealdehyde
Caution: for Research Use Only
Formula:
C₁₅H₁₀O Melting Pt: 103…105 °C Flash Pt: 269.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001254 CAS Number: 642-31-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...