8-Iodoquinoline 97%
Supplier: Ambeed
|
Warning
|
A514192-1G
A514192-100MG
A514192-250MG
103828-438EA
29
USD
103828-438
103839-542
76843-528
77017-346
77017-348
8-Iodoquinoline 97%
8-Iodoquinoline
Caution: for Research Use Only
Formula:
C₉H₆IN Boiling Pt: 320.7 °C (760 mm Hg) Flash Pt: 147.8 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08062396 CAS Number: 1006-47-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...