7-Hydroxy-1-tetralone 98%

Supplier: Ambeed
Warning

A187539-100G A187539-10G A187539-250MG A187539-1G A187539-25G A187539-5G
103816-024EA 6 USD
103816-024 103846-614 76690-748 76690-750 76690-752 77084-310 77084-312 77084-314
7-Hydroxy-1-tetralone 98%
7-Hydroxy-1-tetralone

Caution: for Research Use Only

Formula: C₁₀H₁₀O₂
Boiling Pt: 344.7 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD01312225
CAS Number: 22009-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR