2,6-Diaminopurine 98%

Supplier: Ambeed
Warning

Synonyms: 9H-Purine-2,6-diamine, 7H-Purine-2,6-diamine

A298525-25G A298525-5G A298525-100G A298525-10G A298525-1G A298525-500G
103835-330EA 10 USD
103835-330 76838-028 76838-030 77289-918 77289-920 77289-922 77748-579
2,6-Diaminopurine 98%
2,6-Diaminopurine

Caution: for Research Use Only

Formula: C₅H₆N₆
MW: 150.14 g/mol
Boiling Pt: 749 °C (760 mm Hg)
Melting Pt: 117…122 °C
Flash Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00213668
CAS Number: 1904-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR