7-Azabicyclo[2.2.1]heptane hydrochloride 96%

Supplier: Ambeed
Warning

Synonyms: 7-azabicyclo[2.2.1]heptane hydrochloride

A294335-100MG A294335-250MG A294335-25G A294335-1G A294335-5G
103814-006EA 28 USD
103814-006 103835-146 76693-292 76693-294 76693-296 77305-648 77305-650
7-Azabicyclo[2.2.1]heptane hydrochloride 96%
7-Azabicyclo[2.2.1]heptane hydrochloride

Caution: for Research Use Only

Formula: C₆H₁₂ClN
MW: 133.6200 g/mol
Boiling Pt: 144.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00154183
CAS Number: 27514-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR