6-Chlorouracil 98%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-2,4-dihydroxypyrimidine, 6-Chloropyrimidine-2,4(1H,3H)-dione, 4-Chlorouracil, 4-(3-Bromophenyl)pyrazole, 4-Chloro-2,6-dihydroxypyrimidine, 6-Chloro-2,4(1H,3H)-pyrimidinedione

A149963-100G A149963-500G A149963-10G
103808-734EA 424 USD
103808-734 103842-240 77286-092 77286-094 77317-754 77615-210
6-Chlorouracil 98%
6-Chlorouracil

Caution: for Research Use Only

Formula: C₄H₃ClN₂O₂
Boiling Pt: 300 °C (760 mm Hg)
Melting Pt: 290…295 °C
Storage Temperature: Ambient
MDL Number: MFCD00014595
CAS Number: 4270-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR