5-Methyl-2-thenoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Methylthiophene-2-carboxylic acid

A116820-500G A116820-25G
103824-664EA 8 USD
103824-664 77284-566 77284-568 77571-018
5-Methyl-2-thenoic acid 98%
5-Methyl-2-thenoic acid

Caution: for Research Use Only

Formula: C₆H₆O₂S
Boiling Pt: 274.7 °C (760 mm Hg)
Melting Pt: 135…138 °C
Flash Pt: 120 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005439
CAS Number: 1918-79-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR