Thymine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Methylpyrimidine-2,4(1H,3H)-dione, 5-Methyluracil
A178972-1000G
A178972-25G
A178972-100G
A178972-10G
A178972-5G
A178972-500G
103809-120EA
87
USD
103809-120
103845-728
77083-172
77083-174
77287-100
77287-102
77301-590
77615-508
Thymine 97%
Thymine
Caution: for Research Use Only
Formula:
C₅H₆N₂O₂ MW: 126.11 g/mol Boiling Pt: 389 Melting Pt: 218 Flash Pt: 198 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006026 CAS Number: 65-71-4 Merck Index: 12,09539 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...