5-Methoxy-1H-indole-3-carbaldehyde 97%
Supplier: Ambeed
|
Warning
|
A177468-10G
A177468-1G
A177468-25G
A177468-5G
103845-566EA
16
USD
103845-566
76769-444
76769-446
77082-956
77082-958
5-Methoxy-1H-indole-3-carbaldehyde 97%
5-Methoxy-1H-indole-3-carbaldehyde
Caution: for Research Use Only
Formula:
C10H9NO2 Melting Pt: 179…183 °C Flash Pt: 180.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005623 CAS Number: 10601-19-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...