5,6,7,8-Tetrahydro-1,6-naphthyridine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 5,6,7,8-Tetrahydro-1,6-naphthyridine hcl

A335553-100MG A335553-250MG
103825-692EA 105 USD
103825-692 103836-664 76876-076 76876-078
5,6,7,8-Tetrahydro-1,6-naphthyridine hydrochloride 95%
5,6,7,8-Tetrahydro-1,6-naphthyridine hydrochloride

Caution: for Research Use Only

Formula: C₈H₁₁ClN₂
Storage Temperature: Ambient
MDL Number: MFCD11846187
CAS Number: 1187830-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR