meso-Tetraphenylporphine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5,10,15,20-Tetraphenyl-21H,23H-porphine, 5,10,15,20-Tetraphenylporphyrin, meso-Tetraphenylporphine, TPP
A141389-100G
A141389-10G
A141389-250MG
A141389-1G
A141389-25G
A141389-5G
103802-988EA
10
USD
103802-988
103834-024
76782-942
76782-944
76782-946
77299-210
77299-212
77299-214
meso-Tetraphenylporphine 97%
meso-Tetraphenylporphine
Caution: for Research Use Only
Formula:
C₄₄H₃₀N₄ MW: 614.75 g/mol Melting Pt: 449…451 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011680 CAS Number: 917-23-7 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...