4-Methoxypyridine 98%

Supplier: Ambeed
Warning

A102780-100G A102780-10G A102780-1G A102780-25G A102780-500G A102780-5G
103822-122EA 6 USD
103822-122 77107-268 77107-270 77107-272 77107-274 77107-276 77308-994
4-Methoxypyridine 98%
4-Methoxypyridine

Caution: for Research Use Only

Formula: C₆H₇NO
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00674049
CAS Number: 620-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR